Learn More
Abnova™ Human HOXD8 Full-length ORF (AAH90853.1, 1 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH90853.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3234 |
Molecular Weight (g/mol) | 58.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16198997
|
Abnova™
H00003234-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16188997
|
Abnova™
H00003234-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function. [provided by RefSeq]
Sequence: MSSYFVNPLYSKYKAAAAAAAAAGEAINPTYYDCHFAPGVGGRHAAAAAALQLYGNSAAGFPHAPPQAHAHPHPSPPPSGTGCGGREGRGQEYFHPGGGSPAAAYQAAPPPPPHPPPPPPPPPCGGIACHGEPAKFYGYDNLQRQPIFTTQQEAELVQYPDCKSSSGNIGEDPDHLNQSSSPSQMFPWMRPQAPGRRRGRQTYSRFQTLELEKEFLFNPYLTRKRRIEVSHALALTERQVKIWFQNRRMKWKKENNKDKFPVSRQEVKDGETKKEAQELEEDRAEGLTNSpecifications
AAH90853.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSSYFVNPLYSKYKAAAAAAAAAGEAINPTYYDCHFAPGVGGRHAAAAAALQLYGNSAAGFPHAPPQAHAHPHPSPPPSGTGCGGREGRGQEYFHPGGGSPAAAYQAAPPPPPHPPPPPPPPPCGGIACHGEPAKFYGYDNLQRQPIFTTQQEAELVQYPDCKSSSGNIGEDPDHLNQSSSPSQMFPWMRPQAPGRRRGRQTYSRFQTLELEKEFLFNPYLTRKRRIEVSHALALTERQVKIWFQNRRMKWKKENNKDKFPVSRQEVKDGETKKEAQELEEDRAEGLTN | |
RUO | |
HOXD8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3234 | |
HOXD8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX4/HOX4E/HOX5.4 | |
HOXD8 | |
Recombinant | |
wheat germ expression system |