Learn More
Abnova™ Human HOXA13 Partial ORF (NP_000513, 208 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_000513 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3209 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16196301
|
Abnova™
H00003209-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16186301
|
Abnova™
H00003209-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Expansion of a polyalanine tract in the encoded protein can cause hand-foot-uterus syndrome, also known as hand-foot-genital syndrome. [provided by RefSeq]
Sequence: DKYMDTAGPAAEEFSSRAKEFAFYHQGYAAGPYHHHQPMPGYLDMPVVPGLGGPGESRHEPLGLPMESYQPWALPNGWNGQMYCPKEQAQPPHLWKSTLSpecifications
NP_000513 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX1/HOX1J | |
HOXA13 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3209 | |
HOXA13 (Human) Recombinant Protein (Q01) | |
DKYMDTAGPAAEEFSSRAKEFAFYHQGYAAGPYHHHQPMPGYLDMPVVPGLGGPGESRHEPLGLPMESYQPWALPNGWNGQMYCPKEQAQPPHLWKSTL | |
RUO | |
HOXA13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |