missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HES6 Full-length ORF (NP_061115.2, 1 a.a. - 224 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_061115.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55502 |
Molecular Weight (g/mol) | 50.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121503
|
Abnova™
H00055502-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131503
|
Abnova™
H00055502-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of a subfamily of basic helix-loop-helix transcription repressors that have homology to the Drosophila enhancer of split genes. Members of this gene family regulate cell differentiation in numerous cell types. The protein encoded by this gene functions as a cofactor, interacting with other transcription factors through a tetrapeptide domain in its C-terminus. Alternatively spliced transcript variants encoding different isoforms have been described
Sequence: MAPPAAPGRDRVGREDEDGWETRGDRKARKPLVEKKRRARINESLQELRLLLAGAEVQAKLENAEVLELTVRRVQGVLRGRAREREQLQAEASERFAAGYIQCMHEVHTFVSTCQAIDATVAAELLNHLLESMPLREGSSFQDLLGDALAGPPRAPGRSGWPAGGAPGSPIPSPPGPGDDLCSDLEEAPEAELSQAPAEGPDLVPAALGSLTTAQIARSVWRPWSpecifications
NP_061115.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAPPAAPGRDRVGREDEDGWETRGDRKARKPLVEKKRRARINESLQELRLLLAGAEVQAKLENAEVLELTVRRVQGVLRGRAREREQLQAEASERFAAGYIQCMHEVHTFVSTCQAIDATVAAELLNHLLESMPLREGSSFQDLLGDALAGPPRAPGRSGWPAGGAPGSPIPSPPGPGDDLCSDLEEAPEAELSQAPAEGPDLVPAALGSLTTAQIARSVWRPW | |
RUO | |
HES6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55502 | |
HES6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HES-6/bHLHb41/bHLHc23 | |
HES6 | |
Recombinant | |
wheat germ expression system |