Learn More
R&D Systems™ Human gp130 Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Specifications
Specifications
| Product Type | Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform |
| Quantity | 1 Kit |
| Species | Human |
| Binding Target | gp130/CD130 |
| Gene ID (Entrez) | 3572 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |