Learn More
Abnova™ Human GORASP1 Full-length ORF (NP_114105.1, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_114105.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64689 |
Molecular Weight (g/mol) | 72.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193663
|
Abnova™
H00064689-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183663
|
Abnova™
H00064689-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a membrane protein involved in establishing the stacked structure of the Golgi apparatus. It is a caspase-3 substrate, and cleavage of this encoded protein contributes to Golgi fragmentation in apoptosis. This encoded protein can form a complex with the Golgi matrix protein GOLGA2, and this complex binds to the vesicle docking protein p115. Several alternatively spliced transcript variants of this gene have been identified, but their full-length natures have not been determined. [provided by RefSeq]
Sequence: MGLGVSAEQPAGGAEGFHLHGVQENSPAQQAGLEPYFDFIITIGHSRLNKENDTLKALLKANVEKPVKLEVFNMKTMRVREVEVVPSNMWGGQGLLGASVRFCSFRRASEQVWHVLDVEPSSPAALAGLRPYTDYVVGSDQILQESEDFFTLIESHEGKPLKLMVYNSKSDSCREVTVTPNAAWGGEGSLGCGIGYGYLHRIPTQPPSYHKKPPGTPPPSALPLGAPPPDALPPGPTPEDSPSLETGSRQSDYMEALLQAPGSSMEDPLPGPGSPSHSAPDPDGLPHFMETPLQPPPPVQRVMDPGFLDVSGISLLDNSNASVWPSLPSSTELTTTAVSTSGPEDICSSSSSHERGGEATWSGSEFEVSFLDSPGAQAQADHLPQLTLPDSLTSAASPEDGLSAELLEAQAEEEPASTEGLDTGTEAEGLDSQAQISTTESpecifications
NP_114105.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGLGVSAEQPAGGAEGFHLHGVQENSPAQQAGLEPYFDFIITIGHSRLNKENDTLKALLKANVEKPVKLEVFNMKTMRVREVEVVPSNMWGGQGLLGASVRFCSFRRASEQVWHVLDVEPSSPAALAGLRPYTDYVVGSDQILQESEDFFTLIESHEGKPLKLMVYNSKSDSCREVTVTPNAAWGGEGSLGCGIGYGYLHRIPTQPPSYHKKPPGTPPPSALPLGAPPPDALPPGPTPEDSPSLETGSRQSDYMEALLQAPGSSMEDPLPGPGSPSHSAPDPDGLPHFMETPLQPPPPVQRVMDPGFLDVSGISLLDNSNASVWPSLPSSTELTTTAVSTSGPEDICSSSSSHERGGEATWSGSEFEVSFLDSPGAQAQADHLPQLTLPDSLTSAASPEDGLSAELLEAQAEEEPASTEGLDTGTEAEGLDSQAQISTTE | |
RUO | |
GORASP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64689 | |
GORASP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23443/GOLPH5/GRASP65/MGC118894/MGC118897/P65 | |
GORASP1 | |
Recombinant | |
wheat germ expression system |