missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GIMAP2 Partial ORF (NP_056475.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Spécification
Accession Number | NP_056475.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26157 |
Molecular Weight (g/mol) | 36.74kDa |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16115346
|
Abnova™
H00026157-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125346
|
Abnova™
H00026157-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1. [provided by RefSeq]
Sequence: MDQNEHSHWGPHAKGQCASRSELRIILVGKTGTGKSAAGNSILRKQAFESKLGSQTLTKTCSKSQGSWGNREIVIIDTPDMFSWKDHCEALYKEVQRCYLSpécification
NP_056475.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586D0824/HIMAP2/IMAP2/MGC24275 | |
GIMAP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26157 | |
GIMAP2 (Human) Recombinant Protein (Q01) | |
MDQNEHSHWGPHAKGQCASRSELRIILVGKTGTGKSAAGNSILRKQAFESKLGSQTLTKTCSKSQGSWGNREIVIIDTPDMFSWKDHCEALYKEVQRCYL | |
RUO | |
GIMAP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |