Learn More
Abnova™ Human GALNT1 Partial ORF (NP_065207, 42 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_065207 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2589 |
Molecular Weight (g/mol) | 34.76kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138484
|
Abnova™
H00002589-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16148484
|
Abnova™
H00002589-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. Transcript variants derived from this gene that utilize alternative polyA signals have been described in the literature. [provided by RefSeq]
Sequence: LPAGDVLEPVQKPHEGPGEMGKPVVIPKEDQEKMKEMFKINQFNLMASEMIALNRSLPDVRLEGCKTKVYPDNLPTTSVVIVSpecifications
NP_065207 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GALNAC-T1 | |
GALNT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2589 | |
GALNT1 (Human) Recombinant Protein (Q01) | |
LPAGDVLEPVQKPHEGPGEMGKPVVIPKEDQEKMKEMFKINQFNLMASEMIALNRSLPDVRLEGCKTKVYPDNLPTTSVVIV | |
RUO | |
GALNT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |