Learn More
Abnova™ Human FUK Partial ORF (NP_659496, 169 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_659496 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 197258 |
Molecular Weight (g/mol) | 37.95kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102367
|
Abnova™
H00197258-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16192357
|
Abnova™
H00197258-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date. [provided by RefSeq]
Sequence: RGARVIALPGSPAYAQNHGVYLTDPQGLVLDIYYQGTEAEIQRCVRPDGRVPLVSGVVFFSVETAERLLATHVSPPLDACTYLGLDSGARPVQLSLFFDILHCMAENVTRESpecifications
NP_659496 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
1110046B12Rik/FLJ39408 | |
FUK | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
197258 | |
FUK (Human) Recombinant Protein (Q01) | |
RGARVIALPGSPAYAQNHGVYLTDPQGLVLDIYYQGTEAEIQRCVRPDGRVPLVSGVVFFSVETAERLLATHVSPPLDACTYLGLDSGARPVQLSLFFDILHCMAENVTRE | |
RUO | |
FUK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |