Learn More
ACROBiosystems Human Flt-3 Ligand Protein, His Tag
5297.56 NOK - 38899.20 NOK
Specifications
Accession Number | NP_001450.2 |
---|---|
Molecular Weight (g/mol) | 19.9 kDa |
Source | Human Flt-3 Ligand, His Tag (FLL-H55H7) is expressed from Baculovirus-Insect cells. It contains AA Thr 27 - Pro 185 (Accession # P49771-1). |
Immunogen | Thr 27 - Pro 185 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16457549
|
ACROBiosystems
FLL-H55H7-1MG |
1 mg |
38899.20 NOK
1mg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16447549
|
ACROBiosystems
FLL-H55H7-100UG |
100 μg |
5297.56 NOK
100µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FMS-like tyrosine kinase 3 ligand (Flt-3 Ligand) is also known as FL, Flt3L and FLT3LG, is an α-helical cytokine that promotes the differentiation of multiple hematopoietic cell lineages. FLT3LG is expressed as a noncovalentlylinked dimer by T cells and bone marrow and thymic fibroblasts. Each 36 kDa chain carries approximately 12 kDa of N- and O- linked carbohydrates. FLT3LG is structurally homologous to stem cell factor (SCF) and colony stimulating facor 1 (CSF-1). FLT3LG acts as a growth factor that increases the number of immune cells by activating the hematopoietic progenitors. It also induces the mobilization of the hematopoietic progenitors and stem cells in vivo which may help the system to kill cancer cells. FLT3LG induces the expansion of monocytes and immature dendritic cells as well as early B cell lineage differentiation. FLT3LG cooperates with IL2, IL6, IL7, and IL15 to induce NK cell development and with IL3, IL7 and IL11 to induce terminal B cell maturation. Animal studies also show FLT3LG to reduce the severity of experimentally induced allergic inflammation. FLT3LG is crucial for steady-state pDC and cDC development. A lack of FLT3L results in low levels of DCs.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 22-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_001450.2 | |
Human Flt-3 Ligand, His Tag (FLL-H55H7) is expressed from Baculovirus-Insect cells. It contains AA Thr 27 - Pro 185 (Accession # P49771-1). | |
-20°C | |
Flt-3 Ligand | |
Human | |
PBS, pH7.4 | |
InsectCells | |
Cytokines and Growth Factors |
19.9 kDa | |
Thr 27 - Pro 185 | |
FLT3LG,FL,FLT3L,Flt3 ligand | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |