missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FCRL5 Partial ORF (NP_112571.1, 105 a.a. - 214 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_112571.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 83416 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16150457
|
Abnova™
H00083416-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140457
|
Abnova™
H00083416-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence: LILQAPLSVFEGDSVVLRCRAKAEVTLNNTIYKNDNVLAFLNKRTDFHIPHACLKDNGAYRCTGYKESCCPVSSNTVKIQVQEPFTRPVLRASSFQPISGNPVTLTCETQSpecifications
NP_112571.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BXMAS1/CD307/DKFZp667E2019/DKFZp667F216/FCRH5/FLJ00333/FLJ00397/IRTA2/MGC119590/MGC119592/MGC119593/PRO820 | |
FCRL5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
83416 | |
FCRL5 (Human) Recombinant Protein (Q01) | |
LILQAPLSVFEGDSVVLRCRAKAEVTLNNTIYKNDNVLAFLNKRTDFHIPHACLKDNGAYRCTGYKESCCPVSSNTVKIQVQEPFTRPVLRASSFQPISGNPVTLTCETQ | |
RUO | |
FCRL5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |