Learn More
Invitrogen™ eBioscience™ Human Fc epsilon RI Platinum ELISA Kit
For the detection of human Fc epsilon RI
Brand:  Invitrogen™ BMS2101TEN
Additional Details : Weight : 0.47500kg
 View more versions of this product
Product Code. 15591657
  • 30370.00 NOK / Pack of 10
Estimated Shipment
19-08-2020
Add to basket
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human Fc epsilon RI
Detection antibody (chimeric IgE)
HRP-Conjugate anti-human IgE polyclonal antibody
Human Fc epsilon RI Standard lyophilized, 10ng/mL upon reconstitution
Assay Buffer Concentrate 1x
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
ELISA
Serum, plasma (EDTA), plasma (citrate) and plasma (heparin)
FCER1, FceRI, Fc epsilon RI
Human
0.068ng/mL
Fc epsilon Receptor 1 alpha (FceRI)
10 x 96 Tests