Learn More
Abnova™ Human ESPL1 Partial ORF (NP_036423, 586 a.a. - 701 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_036423 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9700 |
Molecular Weight (g/mol) | 38.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16151176
|
Abnova™
H00009700-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16141176
|
Abnova™
H00009700-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
Stable cohesion between sister chromatids before anaphase and their timely separation during anaphase are critical for chromosome inheritance. In vertebrates, sister chromatid cohesion is released in 2 steps via distinct mechanisms. The first step involves phosphorylation of STAG1 (MIM 604358) or STAG2 (MIM 604359) in the cohesin complex. The second step involves cleavage of the cohesin subunit SCC1 (RAD21; MIM 606462) by ESPL1, or separase, which initiates the final separation of sister chromatids (Sun et al., 2009 [PubMed 19345191]).[supplied by OMIM]
Sequence: REELQAYKAVRADTGQERFNIICDLLELSPEETPAGAWARATHLVELAQVLCYHDFTQQTNCSALDAIREALQLLDSVRPEAQARDQLLDDKAQALLWLYICTLEAKIQEGIERDRSpecifica
NP_036423 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.5kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ESP1/FLJ46492/KIAA0165/SEPARASE/SEPARIN | |
ESPL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9700 | |
ESPL1 (Human) Recombinant Protein (Q01) | |
REELQAYKAVRADTGQERFNIICDLLELSPEETPAGAWARATHLVELAQVLCYHDFTQQTNCSALDAIREALQLLDSVRPEAQARDQLLDDKAQALLWLYICTLEAKIQEGIERDR | |
RUO | |
ESPL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |