Learn More
Abnova™ Human EPB41L1 Full-length ORF (NP_818932.1, 1 a.a. - 779 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_818932.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2036 |
Molecular Weight (g/mol) | 114kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127467
|
Abnova™
H00002036-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16117467
|
Abnova™
H00002036-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane. The protein encoded by this gene is a neuronally-enriched protein that is structurally similar to EPB41. The encoded protein binds and stabilizes D2 and D3 dopamine receptors at the neuronal plasma membrane. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq]
Sequence: MEEKDYSEADGLSERTTPSKAQKSPQKIAKKYKSAICRVTLLDASEYECEVEKHGRGQVLFDLVCEHLNLLEKDYFGLTFCDADSQKNWLDPSKEIKKQIRSSPWNFAFTVKFYPPDPAQLTEDITRYYLCLQLRADIITGRLPCSFVTHALLGSYAVQAELGDYDAEEHVGNYVSELRFAPNQTRELEERIMELHKTYRGMTPGEAEIHFLENAKKLSMYGVDLHHAKDSEGIDIMLGVCANGLLIYRDRLRINRFAWPKILKISYKRSNFYIKIRPGEYEQFESTIGFKLPNHRSAKRLWKVCIEHHTFFRLVSPEPPPKGFLVMGSKFRYSGRTQAQTRQASALIDRPAPFFERSSSKRYTMSRSLDGAEFSRPASVSENHDAGPDGDKRDEDGESGGQRSEAEEGEVRTPTKIKELKFLDKPEDVLLKHQASINELKRTLKEPNSKLIHRDRDWERERRLPSSPASPSPKGTPEKANERAGLREGSEEKVKPPRPRAPESDTGDEDQDQERDTVFLKDNHLAIERKCSSITVSSTSSLEAEVDFTVIGDYHGSAFEDFSRSLPELDRDKSDSDTEGLLFSRDLNKGAPSQDDESGGIEDSPDRGACSTPDMPQFEPVKTETMTVSSLAIRKKIEPEAVLQTRVSAMDNTQENSLKSGKGAAAMIPGPQTVATEIRSLSPIIGKDVLTSTYGATAETLSTSTTTHVTKTVKGGFSETRIEKRIIITGDEDVDQDQALALAIKEAKLQHPDMLVTKAVVYRETDPSPEERDKKPQESSpecifications
NP_818932.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEEKDYSEADGLSERTTPSKAQKSPQKIAKKYKSAICRVTLLDASEYECEVEKHGRGQVLFDLVCEHLNLLEKDYFGLTFCDADSQKNWLDPSKEIKKQIRSSPWNFAFTVKFYPPDPAQLTEDITRYYLCLQLRADIITGRLPCSFVTHALLGSYAVQAELGDYDAEEHVGNYVSELRFAPNQTRELEERIMELHKTYRGMTPGEAEIHFLENAKKLSMYGVDLHHAKDSEGIDIMLGVCANGLLIYRDRLRINRFAWPKILKISYKRSNFYIKIRPGEYEQFESTIGFKLPNHRSAKRLWKVCIEHHTFFRLVSPEPPPKGFLVMGSKFRYSGRTQAQTRQASALIDRPAPFFERSSSKRYTMSRSLDGAEFSRPASVSENHDAGPDGDKRDEDGESGGQRSEAEEGEVRTPTKIKELKFLDKPEDVLLKHQASINELKRTLKEPNSKLIHRDRDWERERRLPSSPASPSPKGTPEKANERAGLREGSEEKVKPPRPRAPESDTGDEDQDQERDTVFLKDNHLAIERKCSSITVSSTSSLEAEVDFTVIGDYHGSAFEDFSRSLPELDRDKSDSDTEGLLFSRDLNKGAPSQDDESGGIEDSPDRGACSTPDMPQFEPVKTETMTVSSLAIRKKIEPEAVLQTRVSAMDNTQENSLKSGKGAAAMIPGPQTVATEIRSLSPIIGKDVLTSTYGATAETLSTSTTTHVTKTVKGGFSETRIEKRIIITGDEDVDQDQALALAIKEAKLQHPDMLVTKAVVYRETDPSPEERDKKPQES | |
RUO | |
EPB41L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2036 | |
EPB41L1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
4.1N/DKFZp686H17242/KIAA0338/MGC11072 | |
EPB41L1 | |
Recombinant | |
wheat germ expression system |