Learn More
Abnova™ Human ENOX2 Partial ORF (NP_872114, 309 a.a. - 379 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010495-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a growth-related cell surface protein. It was identifed because it reacts with the monoclonal antibody KI in cells, such as the ovarian carcinoma line OVCAR-3, also expressing the CAKI surface glycoprotein. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The two activities alternate with a period length of about 24 minutes. The encoded protein also displays prion-like properties. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: AHEKDMEEAKEKFKQALSGILIQFEQIVAVYHSASKQKAWDHFTKAQRKNISVWCKQAEEIRNIHNDELMGSpecifications
NP_872114 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.55kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AHEKDMEEAKEKFKQALSGILIQFEQIVAVYHSASKQKAWDHFTKAQRKNISVWCKQAEEIRNIHNDELMG | |
RUO | |
ENOX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10495 | |
ENOX2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APK1/COVA1/tNOX | |
ENOX2 | |
Recombinant | |
wheat germ expression system |