missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EIF4EBP3 Partial ORF (NP_003723, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003723 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8637 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188925
|
Abnova™
H00008637-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178925
|
Abnova™
H00008637-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the EIF4EBP family which derives it name from proteins that bind to eukaryotic initiation factor 4E and that prevent its assembly into EIF4F. Co-transcription of this gene and the neighboring upstream gene (MASK) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments. [provided by RefSeq]
Sequence: MSTSTSCPIPGGRDQLPDCYSTTPGGTLYATTPGGTRIIYDRKFLLECKNSPIARTPPCCLPQIPGVTTPPTAPLSKLEELKEQETEEEIPDDAQFEMDISpecifications
NP_003723 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
4E-BP3 | |
EIF4EBP3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8637 | |
EIF4EBP3 (Human) Recombinant Protein (Q01) | |
MSTSTSCPIPGGRDQLPDCYSTTPGGTLYATTPGGTRIIYDRKFLLECKNSPIARTPPCCLPQIPGVTTPPTAPLSKLEELKEQETEEEIPDDAQFEMDI | |
RUO | |
EIF4EBP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |