missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EIF3S1 Partial ORF (NP_003749, 157 a.a. - 258 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003749 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8669 |
Molecular Weight (g/mol) | 36.96kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16199005
|
Abnova™
H00008669-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16189005
|
Abnova™
H00008669-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004 [PubMed 14688252]; Fraser et al., 2007 [PubMed 17588516]).[supplied by OMIM]
Sequence: TEFGKLLKDKITQYEKSLYYASFLEVLVRDVCISLEIDDLKKITNSLTVLCSEKQKQEKQSKAKKKKKGVVPGGGLKATMKDDLADYGGYDGGYVQDYEDFMSpecifications
NP_003749 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EIF3S1/eIF3-alpha/eIF3-p35 | |
EIF3J | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8669 | |
EIF3S1 (Human) Recombinant Protein (Q01) | |
TEFGKLLKDKITQYEKSLYYASFLEVLVRDVCISLEIDDLKKITNSLTVLCSEKQKQEKQSKAKKKKKGVVPGGGLKATMKDDLADYGGYDGGYVQDYEDFM | |
RUO | |
EIF3J | |
Wheat Germ (in vitro) | |
GST | |
Liquid |