Learn More
Abnova™ Human ECT2 Partial ORF (AAH06838.1, 46 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001894-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis. [provided by RefSeq]
Sequence: LPKENWLKMLCRHVANTICKADAENLIYTADPESFEVNTKDMDSTLSRASRAIKKTSKKVTRAFSFSKTPKRALRRALMTSHGSVEGRSPSSNDKHVMSRSpecifications
AAH06838.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
LPKENWLKMLCRHVANTICKADAENLIYTADPESFEVNTKDMDSTLSRASRAIKKTSKKVTRAFSFSKTPKRALRRALMTSHGSVEGRSPSSNDKHVMSR | |
RUO | |
ECT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1894 | |
ECT2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10461/MGC138291 | |
ECT2 | |
Recombinant | |
wheat germ expression system |