Learn More
Abnova™ Human DST Partial ORF (NP_899236, 401 a.a. - 500 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_899236 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 667 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16142331
|
Abnova™
H00000667-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16132331
|
Abnova™
H00000667-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the plakin protein family of adhesion junction plaque proteins. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the full-length nature of some variants has not been defined. It has been known that some isoforms are expressed in neural and muscle tissue, anchoring neural intermediate filaments to the actin cytoskeleton, and some isoforms are expressed in epithelial tissue, anchoring keratin-containing intermediate filaments to hemidesmosomes. Consistent with the expression, mice defective for this gene show skin blistering and neurodegeneration. [provided by RefSeq]
Sequence: EDKLILAGNALQSDSKRLESGVQFQNEAEIAGYILECENLLRQHVIDVQILIDGKYYQADQLVQRVAKLRDEIMALRNECSSVYSKGRILTTEQTKLMISSpecifications
NP_899236 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BP240/BPA/BPAG1/CATX-15/D6S1101/DKFZp564B2416/DMH/DT/FLJ46791/KIAA0465/KIAA1470/MACF2 | |
DST | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
667 | |
DST (Human) Recombinant Protein (Q01) | |
EDKLILAGNALQSDSKRLESGVQFQNEAEIAGYILECENLLRQHVIDVQILIDGKYYQADQLVQRVAKLRDEIMALRNECSSVYSKGRILTTEQTKLMIS | |
RUO | |
DST | |
Wheat Germ (in vitro) | |
GST | |
Liquid |