Learn More
ACROBiosystems Human DcR3 / TNFRSF6B Protein, Fc Tag
4883.84 NOK - 42428.38 NOK
Specifications
Accession Number | AAH17065 |
---|---|
Molecular Weight (g/mol) | 56.2 kDa |
Source | Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession # O95407-1). |
Immunogen | Val 30 - His 300 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15847847
|
ACROBiosystems
TNB-H5255-50UG |
50 μg |
4883.84 NOK
50µg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16448109
|
ACROBiosystems
TNB-H5255-1MG |
1 mg |
42428.38 NOK
1mg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Tumor necrosis factor receptor superfamily member 6B (TNFRSF6B) is also known as Decoy Receptor 3 (DcR3), Decoy receptor for Fas ligand and M68, which belongs to the tumor necrosis factor receptor superfamily. TNFRSF6 is a soluble protein which contains four TNFR-Cys repeats. Unlike most of the other members of TNFR superfamily, TNFRSF6 is a soluble protein which contains no transmembrane domain. TNFRSF6B acts as a decoy receptor that competes with death receptors for ligand binding. The protein is postulated to play a regulatory role in suppressing FasL- and LIGHT-mediated cell death and T cell activation as well as to induce angiogenesis via neutralization of TL1A.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 56.2 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH17065 | |
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession # O95407-1). | |
-20°C | |
DcR3 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Other Recombinant Proteins |
56.2 kDa | |
Val 30 - His 300 | |
TNFRSF6B,DCR3,TR6,M68 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |