Learn More
Abnova™ Human DAB1 Full-length ORF (AAH67445.1, 1 a.a. - 553 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001600-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The laminar organization of multiple neuronal types in the cerebral cortex is required for normal cognitive function. In mice, the disabled-1 gene plays a central role in brain development, directing the migration of cortical neurons past previously formed neurons to reach their proper layer. This gene is similar to disabled-1, and the protein encoded by this gene is thought to be a signal transducer that interacts with protein kinase pathways to regulate neuronal positioning in the developing brain. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined. [provided by RefSeq]
Sequence: MSTETELQVAVKTSAKKDSRKKGQDRSEATLIKRFKGEGVRYKAKLIGIDEVSAARGDKLCQDSMMKLKGVVAGARSKGEHKQKIFLTISFGGIKIFDEKTGALQHHHAVHEISYIAKDITDHRAFGYVCGKEGNHRFVAIKTAQAAEPVILDLRDLFQLIYELKQREELEKKAQKDKQCEQAVYQVPTSQKKEGVYDVPKSQPVSNGYSFEDFEERFAAATPNRNLPTDFDEIFEATKAVTQLELFGDMSTPPDITSPPTPATPGDAFIPSSSQTLPASADVFSSVPFGTAAVPSGYVAMGAVLPSFWGQQPLVQQQMVMGAQPPVAQVMPGAQPIAWGQPGLFPATQQPWPTVAGQFPPAAFMPTQTVMPLPAAMFQGPLTPLATVPGTSDSTRSSPQTDKPRQKMGKETFKDFQMAQPPPVPSRKPDQPSLTCTSEAFSSYFNKVGVAQDTDDCDDFDISQLNLTPVTSTTPSTNSPPTPAPRQSSPSKSSASHASDPTTDDIFEEGFESPSKSEEQEAPDGSQASSNSDPFGEPSGEPSGDNISPQAGSSpecifications
AAH67445.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1600 | |
DAB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTETELQVAVKTSAKKDSRKKGQDRSEATLIKRFKGEGVRYKAKLIGIDEVSAARGDKLCQDSMMKLKGVVAGARSKGEHKQKIFLTISFGGIKIFDEKTGALQHHHAVHEISYIAKDITDHRAFGYVCGKEGNHRFVAIKTAQAAEPVILDLRDLFQLIYELKQREELEKKAQKDKQCEQAVYQVPTSQKKEGVYDVPKSQPVSNGYSFEDFEERFAAATPNRNLPTDFDEIFEATKAVTQLELFGDMSTPPDITSPPTPATPGDAFIPSSSQTLPASADVFSSVPFGTAAVPSGYVAMGAVLPSFWGQQPLVQQQMVMGAQPPVAQVMPGAQPIAWGQPGLFPATQQPWPTVAGQFPPAAFMPTQTVMPLPAAMFQGPLTPLATVPGTSDSTRSSPQTDKPRQKMGKETFKDFQMAQPPPVPSRKPDQPSLTCTSEAFSSYFNKVGVAQDTDDCDDFDISQLNLTPVTSTTPSTNSPPTPAPRQSSPSKSSASHASDPTTDDIFEEGFESPSKSEEQEAPDGSQASSNSDPFGEPSGEPSGDNISPQAGS | |
RUO | |
DAB1 | |
Recombinant | |
wheat germ expression system |