Learn More
Abnova™ Human CYP7B1 Partial ORF (NP_004811, 203 a.a. - 286 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_004811 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9420 |
Molecular Weight (g/mol) | 34.98kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190596
|
Abnova™
H00009420-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180596
|
Abnova™
H00009420-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway of extrahepatic tissues, which converts cholesterol to bile acids. This enzyme likely plays a minor role in total bile acid synthesis, but may also be involved in the development of atherosclerosis, neurosteroid metabolism and sex hormone synthesis. [provided by RefSeq]
Sequence: CDNNKFISELRDDFLKFDDKFAYLVSNIPIELLGNVKSIREKIIKCFSSEKLAKMQGWSEVFQSRQDVLEKYYVHEDLEIGAHHSpecifications
NP_004811 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CBAS3/CP7B/SPG5A | |
CYP7B1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9420 | |
CYP7B1 (Human) Recombinant Protein (Q01) | |
CDNNKFISELRDDFLKFDDKFAYLVSNIPIELLGNVKSIREKIIKCFSSEKLAKMQGWSEVFQSRQDVLEKYYVHEDLEIGAHH | |
RUO | |
CYP7B1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |