Learn More
Abnova™ Human CST2 Full-length ORF (NP_001313.1, 1 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_001313.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1470 |
Molecular Weight (g/mol) | 42.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16123651
|
Abnova™
H00001470-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16113651
|
Abnova™
H00001470-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions, where they appear to provide protective functions. The cystatin locus on chromosome 20 contains the majority of the type 2 cystatin genes and pseudogenes. This gene is located in the cystatin locus and encodes a secreted thiol protease inhibitor found at high levels in saliva, tears and seminal plasma. [provided by RefSeq]
Sequence: MAWPLCTLLLLLATQAVALAWSPQEEDRIIEGGIYDADLNDERVQRALHFVISEYNKATEDEYYRRLLRVLRAREQIVGGVNYFFDIEVGRTICTKSQPNLDTCAFHEQPELQKKQLCSFQIYEVPWEDRMSLVNSRCQEASpecifications
NP_001313.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAWPLCTLLLLLATQAVALAWSPQEEDRIIEGGIYDADLNDERVQRALHFVISEYNKATEDEYYRRLLRVLRAREQIVGGVNYFFDIEVGRTICTKSQPNLDTCAFHEQPELQKKQLCSFQIYEVPWEDRMSLVNSRCQEA | |
RUO | |
CST2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1470 | |
CST2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC71924 | |
CST2 | |
Recombinant | |
wheat germ expression system |