missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cripto-1 His-tag Recombinant Protein
HEK293 cells Recombinant Protein
1552.00 NOK - 2755.00 NOK
Specifications
Accession Number | P13385 |
---|---|
Concentration | 200 μg/mL |
For Use With (Application) | Control, ELISA, Western Blot |
Formulation | Dulbecco's PBS with no preservative; pH 7.2 |
Gene ID (Entrez) | 6997 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17616615
|
Invitrogen™
RP87817 |
10 μg |
1552.00 NOK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17626615
|
Invitrogen™
RP87818 |
25 μg |
2755.00 NOK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.Specifications
P13385 | |
Control, ELISA, Western Blot | |
6997 | |
Human Cripto (aa 31-172) His Tag | |
-20°C | |
CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; Epidermal growth factor-like cripto protein CR1; Tdgf1; TDGF2; teratocarcinoma-derived growth factor; teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR) | |
Human | |
Recombinant | |
>95% |
200 μg/mL | |
Dulbecco's PBS with no preservative; pH 7.2 | |
17.4 kD | |
HEK293 cells | |
RUO | |
TDGF1 | |
Unconjugated | |
Liquid |