missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CRHBP Full-length ORF (NP_001873.2, 1 a.a. - 322 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001393-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136677

  • 4115.00 NOK / 10µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Corticotropin-releasing hormone is a potent stimulator of synthesis and secretion of preopiomelanocortin-derived peptides. Although CRH concentrations in the human peripheral circulation are normally low, they increase throughout pregnancy and fall rapidly after parturition. Maternal plasma CRH probably originates from the placenta. Human plasma contains a CRH-binding protein which inactivates CRH and which may prevent inappropriate pituitary-adrenal stimulation in pregnancy. [provided by RefSeq]

Sequence: MSPNFKLQCHFILIFLTALRGESRYLELREAADYDPFLLFSANLKRELAGEQPYRRALRCLDMLSLQGQFTFTADRPQLHCAAFFISEPEEFITIHYDQVSIDCQGGDFLKVFDGWILKGEKFPSSQDHPLPSAERYIDFCESGLSRRSIRSSQNVAMIFFRVHEPGNGFTLTIKTDPNLFPCNVISQTPNGKFTLVVPHQHRNCSFSIIYPVVIKISDLTLGHVNGLQLKKSSAGCEGIGDFVELLGGTGLDPSKMTPLADLCYPFHGPAQMKVGCDNTVVRMVSSGKHVNRVTFEYRQLEPYELENPNGNSIGEFCLSGL
Specifications

Specifications

NP_001873.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRF-BP/CRFBP
CRHBP
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1393
CRHBP (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSPNFKLQCHFILIFLTALRGESRYLELREAADYDPFLLFSANLKRELAGEQPYRRALRCLDMLSLQGQFTFTADRPQLHCAAFFISEPEEFITIHYDQVSIDCQGGDFLKVFDGWILKGEKFPSSQDHPLPSAERYIDFCESGLSRRSIRSSQNVAMIFFRVHEPGNGFTLTIKTDPNLFPCNVISQTPNGKFTLVVPHQHRNCSFSIIYPVVIKISDLTLGHVNGLQLKKSSAGCEGIGDFVELLGGTGLDPSKMTPLADLCYPFHGPAQMKVGCDNTVVRMVSSGKHVNRVTFEYRQLEPYELENPNGNSIGEFCLSGL
RUO
CRHBP
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers