Learn More
Abnova™ Human CREB3L2 Partial ORF (NP_919047.2, 421 a.a. - 518 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064764-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).[supplied by OMIM]
Sequence: ASVVRSRNLLIYEEHSPPEESSSPGSAGELGGWDRGSSLLRVSGLESRPDVDLPHFIISNETSLEKSVLLELQQHLVSAKLEGNETLKVVELDRRVNTSpecifications
NP_919047.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ASVVRSRNLLIYEEHSPPEESSSPGSAGELGGWDRGSSLLRVSGLESRPDVDLPHFIISNETSLEKSVLLELQQHLVSAKLEGNETLKVVELDRRVNT | |
RUO | |
CREB3L2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64764 | |
CREB3L2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BBF2H7/MGC131709/MGC71006 | |
CREB3L2 | |
Recombinant | |
wheat germ expression system |