missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHAF1A Partial ORF (AAH67093, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH67093 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10036 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16121686
|
Abnova™
H00010036-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16111686
|
Abnova™
H00010036-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B; MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).[supplied by OMIM]
Sequence: CKDRPAFPVKKLIQARLPFKRLNLVPKGKADDMSDDQGTSVQSKSPDLEASLDTLENNCHVGSDIDFRPKLVNGKGPLDNFLRNRIETSIGQSTVIIDLTSpecifications
AAH67093 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAF-1/CAF1/CAF1B/CAF1P150/MGC71229/P150 | |
CHAF1A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10036 | |
CHAF1A (Human) Recombinant Protein (Q01) | |
CKDRPAFPVKKLIQARLPFKRLNLVPKGKADDMSDDQGTSVQSKSPDLEASLDTLENNCHVGSDIDFRPKLVNGKGPLDNFLRNRIETSIGQSTVIIDLT | |
RUO | |
CHAF1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |