missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDS2 Partial ORF (NP_003809, 1 a.a. - 67 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number NP_003809
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8760
Molecular Weight (g/mol) 33.11kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16199175
View Documents Promotion Details
Abnova™
H00008760-Q01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16189175
View Documents Promotion Details
Abnova™
H00008760-Q01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Breakdown products of phosphoinositides are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. This gene encodes an enzyme which regulates the amount of phosphatidylinositol available for signaling by catalyzing the conversion of phosphatidic acid to CDP-diacylglycerol. This enzyme is an integral membrane protein localized to two subcellular domains, the matrix side of the inner mitochondrial membrane where it is thought to be involved in the synthesis of phosphatidylglycerol and cardiolipin and the cytoplasmic side of the endoplasmic reticulum where it functions in phosphatidylinositol biosynthesis. Two genes encoding this enzyme have been identified in humans, one mapping to human chromosome 4q21 and a second to 20p13. [provided by RefSeq]

Sequence: MTELRQRVAHEPVAPPEDKESESEAKVDGETASDSESRAESAPLPVSADDTPEVLNRALSNLSSRWK
Specifications

Specifications

NP_003809
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.11kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38111
CDS2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
8760
CDS2 (Human) Recombinant Protein (Q01)
MTELRQRVAHEPVAPPEDKESESEAKVDGETASDSESRAESAPLPVSADDTPEVLNRALSNLSSRWK
RUO
CDS2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications