Learn More
Abnova™ Human CDH26 Full-length ORF (NP_068582.2, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_068582.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 60437 |
Molecular Weight (g/mol) | 44.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16103213
|
Abnova™
H00060437-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16193203
|
Abnova™
H00060437-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cadherins are a family of adhesion molecules that mediate Ca2+-dependent cell-cell adhesion in all solid tissues and modulate a wide variety of processes, including cell polarization and migration. Cadherin domains occur as repeats in the extracellular region and are thought to contribute to the sorting of heterogeneous cell types and the maintenance of orderly structures such as epithelium. This gene encodes a cadherin domain-containing protein whose specific function has not yet been determined. Alternative splicing occurs at this locus and two transcript variants, encoding distinct proteins, have been identified. [provided by RefSeq]
Sequence: MKPLIWTWSDVEGQRPALLICTAAAGPTQGVKDLEEVPPSAASQSAQARCALGSWGYGKPFEPRSVKNIHSTPAYPDATMHRQLLAPVEGRMAETLNQKLHVANVLEDDPGYLPHVYSEEGECGGAPSLSSLASLEQELQPDLLDSLGSKATPFEEIYSESGVPSSpecifications
NP_068582.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKPLIWTWSDVEGQRPALLICTAAAGPTQGVKDLEEVPPSAASQSAQARCALGSWGYGKPFEPRSVKNIHSTPAYPDATMHRQLLAPVEGRMAETLNQKLHVANVLEDDPGYLPHVYSEEGECGGAPSLSSLASLEQELQPDLLDSLGSKATPFEEIYSESGVPS | |
RUO | |
CDH26 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
60437 | |
CDH26 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VR20 | |
CDH26 | |
Recombinant | |
wheat germ expression system |