missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD27 Ligand (TNFSF7, CD70) Fc Chimera, Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ A42566
Additional Details : Weight : 0.01070kg
Description
The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.Specifications
P32970 | |
protein with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human CD27 Ligand (TNFSF7, CD70) Fc Chimera | |
HEK293 cells | |
RUO | |
CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8 A; tumor necrosis factor ligand superfamily member 7 | |
Human | |
Recombinant | |
≥ 90% |
Control, ELISA, Immunohistochemistry, Western Blot | |
970 | |
50 μg | |
-20°C | |
<1 EU/μg | |
Cd70 | |
Unconjugated | |
Lyophilized |