Learn More
Abnova™ Human CBFA2T3 Full-length ORF (AAH62624.1, 1 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH62624.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 863 |
Molecular Weight (g/mol) | 93.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16182641
|
Abnova™
H00000863-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16192641
|
Abnova™
H00000863-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The t(16;21)(q24;q22) translocation is a rare but recurrent chromosomal abnormality associated with therapy-related myeloid malignancies. The translocation produces a chimeric gene made up of the 5'-region of the AML1 gene fused to the 3'-region of this gene. In addition, this gene is a putative breast tumor suppressor. Two transcript variants encoding different isoforms have been found for this gene, and a brefeldin A-sensitive association of RII-alpha protein with one of the isoforms has been demonstrated in the Golgi apparatus. [provided by RefSeq]
Sequence: MPASRLRDRAASSASGSTCGSMSQTHPVLESGLLASAGCSAPRGPRKGGPAPVDRKAKASAMPDSPAEVKTQPRSTPPSMPPPPPAASQGATRPPSFTPHTLMNGSSHSPTAINGAPCTPNGFSNGPATSSTASLSTQHLPPACGARQLSKLKRFLTTLQQFGSDISPEIGERVRTLVLGLVNSTLTIEEFHSKLQEATNFPLRPFVIPFLKANLPLLQRELLHCARLAKQTPAQYLAQHEQLLLDASASSPIDSSELLLEVNENGKRRTPDRTKENGSDRDPLHPQRYSPSNGPPQPTPPPHYRLEDIAMAHHFRDAYRHPDPRELRERHRPLVVPGSRQEEVIDHKLTEREWAEEWKHLNNLLNCIMDMVEKTRRSLTVLRRCQEADREELNHWARRYSDAEDTKKGPAPAAARPRSSSAGPEGPQLDVPREFLPRTLTGYVPEDIWRKAEEAVNEVKRQAMSELQKAVSDAERKAHELITTERAKMERALAEAKRQASEDALTVINQQEDSSESCWNCGRKASETCSGCNAARYCGSFCQHRDWEKHHHVCGQSLQGPTAVVADPVPGPPEAAHSLGPSLPVGAASPSEAGSAGPSRPGSPSPPGPLDTVPRSpecifications
AAH62624.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
93.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPASRLRDRAASSASGSTCGSMSQTHPVLESGLLASAGCSAPRGPRKGGPAPVDRKAKASAMPDSPAEVKTQPRSTPPSMPPPPPAASQGATRPPSFTPHTLMNGSSHSPTAINGAPCTPNGFSNGPATSSTASLSTQHLPPACGARQLSKLKRFLTTLQQFGSDISPEIGERVRTLVLGLVNSTLTIEEFHSKLQEATNFPLRPFVIPFLKANLPLLQRELLHCARLAKQTPAQYLAQHEQLLLDASASSPIDSSELLLEVNENGKRRTPDRTKENGSDRDPLHPQRYSPSNGPPQPTPPPHYRLEDIAMAHHFRDAYRHPDPRELRERHRPLVVPGSRQEEVIDHKLTEREWAEEWKHLNNLLNCIMDMVEKTRRSLTVLRRCQEADREELNHWARRYSDAEDTKKGPAPAAARPRSSSAGPEGPQLDVPREFLPRTLTGYVPEDIWRKAEEAVNEVKRQAMSELQKAVSDAERKAHELITTERAKMERALAEAKRQASEDALTVINQQEDSSESCWNCGRKASETCSGCNAARYCGSFCQHRDWEKHHHVCGQSLQGPTAVVADPVPGPPEAAHSLGPSLPVGAASPSEAGSAGPSRPGSPSPPGPLDTVPR | |
RUO | |
CBFA2T3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
863 | |
CBFA2T3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETO2/MTG16/MTGR2/ZMYND4 | |
CBFA2T3 | |
Recombinant | |
wheat germ expression system |