missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CAPSL Full-length ORF (AAH17586.2, 1 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number AAH17586.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 133690
Molecular Weight (g/mol) 49.5kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16118013
View Documents Promotion Details
Abnova™
H00133690-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16128013
View Documents Promotion Details
Abnova™
H00133690-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence: MAIQAKKKLTTATNPIERLRLQCLARGSAGIKGLGRVFRIMDDDNNRTLDFKEFMKGLNDYAVVMEKEEVEELFRRFDKDGNGTIDFNEFLLTLRPPMSRARKEVIMQAFRKLDKTGDGVITIEDLREVYNAKHHPKYQNGEWSEEQVFRKFLDNFDSPYDKDGLVTPEEFMNYYAGVSASIDTDVYFIIMMRTAWKL
Specifications

Specifications

AAH17586.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.5kDa
Glutathione Sepharose 4 Fast Flow
MAIQAKKKLTTATNPIERLRLQCLARGSAGIKGLGRVFRIMDDDNNRTLDFKEFMKGLNDYAVVMEKEEVEELFRRFDKDGNGTIDFNEFLLTLRPPMSRARKEVIMQAFRKLDKTGDGVITIEDLREVYNAKHHPKYQNGEWSEEQVFRKFLDNFDSPYDKDGLVTPEEFMNYYAGVSASIDTDVYFIIMMRTAWKL
RUO
CAPSL
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
133690
CAPSL (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC26610
CAPSL
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications