missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CALN1 Partial ORF (NP_113656, 106 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083698-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190517

  • 4115.00 NOK / 10µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein with high similarity to the calcium-binding proteins of the calmodulin family. The encoded protein contains two EF-hand domains and potential calcium-binding sites. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: VSSEGRDGFLGNTIDSIFWQFDMQRITLEELKHILYHAFRDHLTMKDIENIIINEEESLNETSGNCQTEFEGVHSQKQ
Specifications

Specifications

NP_113656
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.32kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VSSEGRDGFLGNTIDSIFWQFDMQRITLEELKHILYHAFRDHLTMKDIENIIINEEESLNETSGNCQTEFEGVHSQKQ
RUO
CALN1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
83698
CALN1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CALN1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers