missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C2orf25 Full-length ORF (NP_056517.1, 1 a.a. - 296 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_056517.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27249 |
Molecular Weight (g/mol) | 59.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188702
|
Abnova™
H00027249-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16198702
|
Abnova™
H00027249-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a mitochondrial protein that is involved in an early step of vitamin B12 metabolism. Vitamin B12 (cobalamin) is essential for normal development and survival in humans. Mutations in this gene cause methylmalonic aciduria and homocystinuria type cblD (MMADHC), a disorder of cobalamin metabolism that is characterized by decreased levels of the coenzymes adenosylcobalamin and methylcobalamin. Pseudogenes have been identified on chromosomes 11 and X
Sequence: MANVLCNRARLVSYLPGFCSLVKRVVNPKAFSTAGSSGSDESHVAAAPPDICSRTVWPDETMGPFGPQDQRFQLPGNIGFDCHLNGTASQKKSLVHKTLPDVLAEPLSSERHEFVMAQYVNEFQGNDAPVEQEINSAETYFESARVECAIQTCPELLRKDFESLFPEVANGKLMILTVTQKTKNDMTVWSEEVEIEREVLLEKFINGAKEICYALRAEGYWADFIDPSSGLAFFGPYTNNTLFETDERYRHLGFSVDDLGCCKVIRHSLWGTHVVVGSIFTNATPDSHIMKKLSGNSpecifications
NP_056517.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MANVLCNRARLVSYLPGFCSLVKRVVNPKAFSTAGSSGSDESHVAAAPPDICSRTVWPDETMGPFGPQDQRFQLPGNIGFDCHLNGTASQKKSLVHKTLPDVLAEPLSSERHEFVMAQYVNEFQGNDAPVEQEINSAETYFESARVECAIQTCPELLRKDFESLFPEVANGKLMILTVTQKTKNDMTVWSEEVEIEREVLLEKFINGAKEICYALRAEGYWADFIDPSSGLAFFGPYTNNTLFETDERYRHLGFSVDDLGCCKVIRHSLWGTHVVVGSIFTNATPDSHIMKKLSGN | |
RUO | |
MMADHC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27249 | |
C2orf25 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C2orf25/CL25022 | |
MMADHC | |
Recombinant | |
wheat germ expression system |