missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C1orf34 Full-length ORF (XP_939747.1, 1 a.a. - 221 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022996-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16126832

  • 3655.00 NOK / 10µg
Estimated Shipment: 19-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MKAAYLSMFGKEDHKPFGDDEVELFRAVPGLKLKIAGKSLPTEKFAIRKSRRYFSSNPISLPVPALEMMYIWNGYAVIGKQPKLTDGILEIITKAEEMLEKGPENEYSVDDECLVKLLKGLCLKYLGRVQEAEENFRSISANEKKIKYDHYLIPNALLELALLLMEQDRNEEAIKLLESAKQNYKNYSMESRTHFRIQAATLQAKSSLENSSRSMVSSVSL
Specifications

Specifications

XP_939747.1
Liquid
22996
C1orf34 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKAAYLSMFGKEDHKPFGDDEVELFRAVPGLKLKIAGKSLPTEKFAIRKSRRYFSSNPISLPVPALEMMYIWNGYAVIGKQPKLTDGILEIITKAEEMLEKGPENEYSVDDECLVKLLKGLCLKYLGRVQEAEENFRSISANEKKIKYDHYLIPNALLELALLLMEQDRNEEAIKLLESAKQNYKNYSMESRTHFRIQAATLQAKSSLENSSRSMVSSVSL
RUO
TTC39A
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf34/DEME-6/KIAA0452
TTC39A
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers