missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C13orf7 Partial ORF (NP_078822, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_078822 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79596 |
Molecular Weight (g/mol) | 36.19kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144243
|
Abnova™
H00079596-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134243
|
Abnova™
H00079596-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MLSHTVRKHLRKTRLELLHKEYEDEIDCLQKEVEELKSKNLSLESQIKTILDPLTLVQGNQNEDKHLVTDNPSKINPETVAEWKKKLRTANEIYESpecifications
NP_078822 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C13orf7/DKFZp686A01276/DKFZp686N15250/DKFZp686O03173/FLJ13449/FLJ25774 | |
RNF219 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79596 | |
C13orf7 (Human) Recombinant Protein (Q01) | |
MLSHTVRKHLRKTRLELLHKEYEDEIDCLQKEVEELKSKNLSLESQIKTILDPLTLVQGNQNEDKHLVTDNPSKINPETVAEWKKKLRTANEIYE | |
RUO | |
RNF219 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |