missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BPY2IP1 Partial ORF (NP_060644.4, 929 a.a. - 1026 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055201-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: SGSASSRPGVSATPPKSPVYLDLAYLPSGSSAHLVDEEFFQRVRALCYVISGQDQRKEEGMRAVLDALLASKQHWDRDLQVTLIPTFDSVAMHTWYAESpecifications
NP_060644.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SGSASSRPGVSATPPKSPVYLDLAYLPSGSSAHLVDEEFFQRVRALCYVISGQDQRKEEGMRAVLDALLASKQHWDRDLQVTLIPTFDSVAMHTWYAE | |
RUO | |
MAP1S | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55201 | |
BPY2IP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BPY2IP1/C19orf5/FLJ10669/MAP8/MGC133087/VCY2IP-1/VCY2IP1 | |
MAP1S | |
Recombinant | |
wheat germ expression system |