missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARSF Full-length ORF (NP_004033.2, 1 a.a. - 590 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000416-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141921

  • 5420.23 NOK / 25µg

Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the sulfatase family, and more specifically, the arylsulfatase subfamily. Members of the subfamily share similarity in sequence and splice sites, and are clustered together on chromosome X, suggesting that they are derived from recent gene duplication events. Sulfatases are essential for the correct composition of bone and cartilage matrix. The activity of this protein, unlike that of arylsulfatase E, is not inhibited by warfarin. [provided by RefSeq]

Sequence: MRPRRPLVFMSLVCALLNTCQAHRVHDDKPNIVLIMVDDLGIGDLGCYGNDTMRTPHIDRLAREGVRLTQHISAASLCSPSRSAFLTGRYPIRSGMVSSGNRRVIQNLAVPAGLPLNETTLAALLKKQGYSTGLIGKWHQGLNCDSRSDQCHHPYNYGFDYYYGMPFTLVDSCWPDPSRNTELAFESQLWLCVQLVAIAILTLTFGKLSGWVSVPWLLIFSMILFIFLLGYAWFSSHTSPLYWDCLLMRGHEITEQPMKAERAGSIMVKEAISFLERHSKETFLLFFSFLHVHTPLPTTDDFTGTSKHGLYGDNVEEMDSMVGKILDAIDDFGLRNNTLVYFTSDHGGHLEARRGHAQLGGWNGIYKGGKGMGGWEGGIRVPGIVRWPGKVPAGRLIKEPTSLMDILPTVASVSGGSLPQDRVIDGRDLMPLLQGNVRHSEHEFLFHYCGSYLHAVRWIPKDDSGSVWKAHYVTPVFQPPASGGCYVTSLCRCFGEQVTYHNPPLLFDLSRDPSESTPLTPATEPLHDFVIKKVANALKEHQETIVPVTYQLSELNQGRTWLKPCCGVFPFCLCDKEEEVSQPRGPNEKR
Specifications

Specifications

NP_004033.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
RUO
ARSF
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
416
ARSF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASF
ARSF
Yes
Videos
SDS
Documents
Certificates