Learn More
Abnova™ Human ARHGEF3 Full-length ORF (AAH99715.1, 1 a.a. - 324 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH99715.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 50650 |
Molecular Weight (g/mol) | 63.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139412
|
Abnova™
H00050650-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16149412
|
Abnova™
H00050650-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Rho-like GTPases are involved in a variety of cellular processes, and they are activated by binding GTP and inactivated by conversion of GTP to GDP by their intrinsic GTPase activity. Guanine nucleotide exchange factors (GEFs) accelerate the GTPase activity of Rho GTPases by catalyzing their release of bound GDP. This gene encodes a guanine nucleotide exchange factor, which specifically activates two members of the Rho GTPase family: RHOA and RHOB, both of which have a role in bone cell biology. It has been identified that genetic variation in this gene plays a role in the determination of bone mineral density (BMD), indicating the implication of this gene in postmenopausal osteoporosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MKLPCLSSYDSYCSNQVAAKALLDHKKQDHRVQDFLQRCLESPFSRKLDLWNFLDIPRSRLVKYPLLLREILRHTPNDNPDQQHLEEAINIIQGIVAEINTKTGESECRYYKERLLYLEEGQKDSLIDSSRVVCCHGELKNNRGVKLHVFLFQEVLVITRAVTHNEQLCYQLYRQPIPVKDLLLEDLQDGEVRLGGSLRGAFSNNERIKNFFRVSFKNGSQSQTHSLQANDTFNKQQWLNCIRQAKETVLCAAGQAGVLDSEGSFLNPTTGSRELQGETKLEQMDQSDSESDCSMDTSEVSLDCERMEQTDSSCGNSRHGESNVSpecifications
AAH99715.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434F2429/FLJ98126/GEF3/MGC118905/STA3/XPLN | |
ARHGEF3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
50650 | |
ARHGEF3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ARHGEF3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |