Learn More
Abnova™ Human ARGFX Full-length ORF (NP_001012677.1, 1 a.a. - 315 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00503582-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the ARGFX homeobox gene family. [provided by RefSeq]
Sequence: MRNRMAPENPQPDPFINRNYSNMKVIPPQDPASPSFTLLSKLECSGTVSAYCSLNLPGSTDPPTSASRVAATTAIRRRHKERTSFTHQQYEELEALFSQTMFPDRNLQEKLALRLDLPESTVKVWFRNRRFKLKKQQQQQSAKQRNQILPSKKNVPTSPRTSPSPYAFSPVISDFYSSLPSQPLDPSNWAWNSTFTESSTSDFQMQDTQWERLVASVPALYSDAYDIFQIIELYNLPDENEISSSSFHCLYQYLSPTKYQVGGQGSSLSIFAGPAVGLSPAQTWPNMTSQAFEAYSLTDSLEFQKTSNMVDLGFLSpecifications
NP_001012677.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARGFX | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
503582 | |
ARGFX (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARGFX | |
Wheat Germ (in vitro) | |
GST | |
Liquid |