missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human APS Partial ORF (NP_066189, 311 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010603-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125402

  • 5540.00 NOK / 25µg
Estimated Shipment: 19-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is expressed in B lymphocytes and contains pleckstrin homology and src homology 2 (SH2) domains. In Burkitt's lymphoma cell lines, it is tyrosine-phosphorylated in response to B cell receptor stimulation. Because it binds Shc independent of stimulation and Grb2 after stimulation, it appears to play a role in signal transduction from the receptor to the Shc/Grb2 pathway. [provided by RefSeq]

Sequence: EEDTELSCTRGGCLASRVASCSCELLTDAVDLPRPPETTAVGAVVTAPHSRGRDAVRESLIHVPLETFLQTLESPGGSGSDSNNTGEQGAETDPEAEPEL
Specifications

Specifications

NP_066189
Liquid
10603
APS (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APS
SH2B2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EEDTELSCTRGGCLASRVASCSCELLTDAVDLPRPPETTAVGAVVTAPHSRGRDAVRESLIHVPLETFLQTLESPGGSGSDSNNTGEQGAETDPEAEPEL
RUO
SH2B2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers