missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AMPD1 Full-length ORF (AAI41601.1, 1 a.a. - 747 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
3655.00 NOK - 5540.00 NOK
Specifications
Accession Number | AAI41601.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 270 |
Molecular Weight (g/mol) | 109.12kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141541
|
Abnova™
H00000270-P01.25UG |
25 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16131541
|
Abnova™
H00000270-P01.10UG |
10 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Adenosine monophosphate deaminase 1 catalyzes the deamination of AMP to IMP in skeletal muscle and plays an important role in the purine nucleotide cycle. Two other genes have been identified, AMPD2 and AMPD3, for the liver- and erythocyte-specific isoforms, respectively. Deficiency of the muscle-specific enzyme is apparently a common cause of exercise-induced myopathy and probably the most common cause of metabolic myopathy in the human. [provided by RefSeq]
Sequence: MPLFKLPAEEKQIDDAMRNFAEKVFASEVKDEGGRQEISPFDVDEICPISHHEMQAHIFHLETLSTSTEARRKKRFQGRKTVNLSIPLSETSSTKLSHIDEYISSSPTYQTVPDFQRVQITGDYASGVTVEDFEIVCKGLYRALCIREKYMQKSFQRFPKTPSKYLRNIDGEAWVANESFYPVFTPPVKKGEDPFRTDNLPENLGYHLKMKDGVVYVYPNEAAVSKDEPKPLPYPNLDTFLDDMNFLLALIAQGPVKTYTHRRLKFLSSKFQVHQMLNEMDELKELKNNPHRDFYNCRKVDTHIHAAACMNQKHLLRFIKKSYQIDADRVVYSTKEKNLTLKELFAKLKMHPYDLTVDSLDVHAGRQTFQRFDKFNDKYNPVGASELRDLYLKTDNYINGEYFATIIKEVGADLVEAKYQHAEPRLSIYGRSPDEWSKLSSWFVCNRIHCPNMTWMIQVPRIYDVFRSKNFLPHFGKMLENIFMPVFEATINPQADPELSVFLKHITGFDSVDDESKHSGHMFSSKSPKPQEWTLEKNPSYTYYAYYMYANIMVLNSLRKERGMNTFLFRPHCGEAGALTHLMTAFMIADDISHGLNLKKSPVLQYLFFLAQIPIAMSPLSNNSLFLEYAKNPFLDFLQKGLMISLSTDDPMQFHFTKEPLMEEYAIAAQVFKLSTCDMCEVARNSVLQCGISHEEKVKFLGDNYLEEGPAGNDIRRTNVAQIRMAYRYETWCYELNLIAEGLKSTESpecifications
AAI41601.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109.12kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAD/MADA | |
AMPD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
270 | |
AMPD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AMPD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |