Learn More
ACROBiosystems Human Alpha-Synuclein Protein, His Tag
2421.74 NOK - 10141.03 NOK
Specifications
Accession Number | NP_000336.1 |
---|---|
Molecular Weight (g/mol) | 16.3 kDa |
Source | Human Alpha-Synuclein, His Tag (ALN-H52H8) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1). |
Immunogen | Met 1 - Ala 140 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17106071
|
ACROBiosystems
ALN-H52H8-200UG |
200 μg |
2421.74 NOK
200µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17196061
|
ACROBiosystems
ALN-H52H8-1MG |
1 mg |
10141.03 NOK
1mg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Neuronal protein that plays several roles in synaptic activity such as regulation of synaptic vesicle trafficking and subsequent neurotransmitter release. Participates as a monomer in synaptic vesicle exocytosis by enhancing vesicle priming, fusion and dilation of exocytotic fusion pores. Mechanistically, acts by increasing local Ca2+ release from microdomains which is essential for the enhancement of ATP-induced exocytosis. Acts also as a molecular chaperone in its multimeric membrane-bound state, assisting in the folding of synaptic fusion components called SNAREs (Soluble NSF Attachment Protein REceptors) at presynaptic plasma membrane in conjunction with cysteine string protein-alpha/DNAJC5. This chaperone activity is important to sustain normal SNARE-complex assembly during aging. Plays also a role in the regulation of the dopamine neurotransmission by associating with the dopamine transporter (DAT1) and thereby modulating its activity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.3 kDa. The protein migrates as 19 kDa under reducing (R) condition (SDS-PAGE).Specifications
NP_000336.1 | |
Human Alpha-Synuclein, His Tag (ALN-H52H8) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1). | |
-20°C | |
Alpha-Synuclein | |
Human | |
100 mM NaAC, pH7.0 | |
E.coli | |
Other Recombinant Proteins |
16.3 kDa | |
Met 1 - Ala 140 | |
SNCA,NACP,PARK1,alpha-Synuclein | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |