missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AK5 Partial ORF (NP_777283, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_777283 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26289 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135466
|
Abnova™
H00026289-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125466
|
Abnova™
H00026289-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the adenylate kinase family, which is involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. This member is related to the UMP/CMP kinase of several species. It is located in the cytosol and expressed exclusively in brain. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MNTNDAKEYLARREIPQLFESLLNGLMCSKPEDPVEYLESCLQKVKELGGCDKVKWDTFVSQEKKTLPPLNGGQSRRSFLRNVMPENSNFPYRRYDRLPPSpecifications
NP_777283 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AK6/MGC33326 | |
AK5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26289 | |
AK5 (Human) Recombinant Protein (Q01) | |
MNTNDAKEYLARREIPQLFESLLNGLMCSKPEDPVEYLESCLQKVKELGGCDKVKWDTFVSQEKKTLPPLNGGQSRRSFLRNVMPENSNFPYRRYDRLPP | |
RUO | |
AK5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |