missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AIP Partial ORF (NP_003968.1, 156 a.a. - 249 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_003968.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9049 |
Molecular Weight (g/mol) | 36.08kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189805
|
Abnova™
H00009049-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16199805
|
Abnova™
H00009049-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus. [provided by RefSeq]
Sequence: KVESPGTYQQDPWAMTDEEKAKAVPLIHQEGNRLYREGHVKEAAAKYYDAIACLKNLQMKEQPGSPEWIQLDKQITPLLLNYCQCKLVVEEYYESpecifications
NP_003968.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARA9/FKBP16/FKBP37/SMTPHN/XAP2 | |
AIP | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9049 | |
AIP (Human) Recombinant Protein (Q01) | |
KVESPGTYQQDPWAMTDEEKAKAVPLIHQEGNRLYREGHVKEAAAKYYDAIACLKNLQMKEQPGSPEWIQLDKQITPLLLNYCQCKLVVEEYYE | |
RUO | |
AIP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |