missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADAMTS6 Partial ORF (NP_922932.2, 725 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_922932.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11174 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16133836
|
Abnova™
H00011174-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16123836
|
Abnova™
H00011174-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. [provided by RefSeq]
Sequence: DSLPRGGYMEVVQIPRGSVHIEVREVAMSKNYIALKSEGDDYYINGAWTIDWPRKFDVAGTAFHYKRPTDEPESLEALGPTSENLIVMVLLQEQNLGIRYSpecifications
NP_922932.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADAM-TS6 | |
ADAMTS6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11174 | |
ADAMTS6 (Human) Recombinant Protein (Q01) | |
DSLPRGGYMEVVQIPRGSVHIEVREVAMSKNYIALKSEGDDYYINGAWTIDWPRKFDVAGTAFHYKRPTDEPESLEALGPTSENLIVMVLLQEQNLGIRY | |
RUO | |
ADAMTS6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |