missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADAMDEC1 Partial ORF (NP_055294, 361 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number NP_055294
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 27299
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16195846
View Documents Promotion Details
Abnova™
H00027299-Q01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16105856
View Documents Promotion Details
Abnova™
H00027299-Q01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This encoded protein is thought to be a secreted protein belonging to the disintegrin metalloproteinase family. Its expression is upregulated during dendritic cells maturation. This protein may play an important role in dendritic cell function and their interactions with germinal center T cells. [provided by RefSeq]

Sequence: PDVPFNTKCPSGSCVMNQYLSSKFPKDFSTSCRAHFERYLLSQKPKCLLQAPIPTNIMTTPVCGNHLLEVGEDCDCGSPKECTNLCCEALTCKLKPGTDCGGDAPNHTTE
Specifications

Specifications

NP_055294
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
M12.219
ADAMDEC1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
27299
ADAMDEC1 (Human) Recombinant Protein (Q01)
PDVPFNTKCPSGSCVMNQYLSSKFPKDFSTSCRAHFERYLLSQKPKCLLQAPIPTNIMTTPVCGNHLLEVGEDCDCGSPKECTNLCCEALTCKLKPGTDCGGDAPNHTTE
RUO
ADAMDEC1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications