missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human 40787 Full-length ORF (AAH12161, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH12161 |
---|---|
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1731 |
Molecular Weight (g/mol) | 66.11kDa |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16194041
|
Abnova™
H00001731-P01.25UG |
25μg |
6245.00 NOK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16184041
|
Abnova™
H00001731-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the septin family of GTPases. Members of this family are required for cytokinesis. This gene encodes a protein associated with the tau-based paired helical filament core, and may contribute to the formation of neurofibrillary tangles in Alzheimer's disease. [provided by RefSeq]
Sequence: MDKEYVGFAALPNQLHRKSVKKGFDFTLMVAGESGLGKSTLINSLFLTNLYEDRQVPEASARLTQTLAIERRGVEIEEGGVKVKLTLVDTPGFGDSVDCSDCWLPVVKFIEEQFEQYLRDESGLNRKNIQDSRVHCCLYFISPFGRGLRPLDVAFLRAVHEKVNIIPVIGKADALMPQETQALKQKIRDQLKEEEIHIYQFPECDSDEDEDFKRQDAEMKESIPFAVVGSCEVVRDGGNRPVRGRRYSWGTVEVENPHHCDFLNLRRMLVQTHLQDLKEVTHDLLYEGYRARRLQSLARPGARDRASRSKLSRQSATEIPLPMLPLADTEKLIREKDEELRRMQEMLEKMQAQMQRSQAQGEQSDALSpecifications
AAH12161 | |
1731 | |
Glutathione Sepharose 4 Fast Flow | |
RUO | |
SEPT1 | |
Recombinant | |
Liquid |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.11kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SEPT1 | |
Wheat Germ (in vitro) | |
wheat germ expression system |