missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70/HSPA1A Antibody (1H11) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-59339JF585
This item is not returnable.
View return policy
Description
HSP70/HSPA1A Monoclonal antibody specifically detects HSP70/HSPA1A in Bovine,Human,Mouse,Porcine,Rat samples. It is validated for Flow Cytometry,Western Blot,Flow (Cell Surface),Immunocytochemistry/ Immunofluorescence
Specifications
| HSP70/HSPA1A | |
| Monoclonal | |
| Janelia Fluor 585 | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
| Human native HSP70 protein | |
| 0.1 mL | |
| Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 3303 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Western Blot, Flow Cytometry (Cell Surface), Immunocytochemistry/Immunofluorescence | |
| 1H11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Human, Mouse, Pig, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction