Learn More
ACROBiosystems HRP-Human Serum Albumin Protein, His Tag (SPR verified)
Product Code. 31453572 Shop All ACROBiosystems Products
Change view
Click to view available options
Quantity:
200 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31453572 200 μg
31452560 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31453572 Supplier ACROBiosystems Supplier No. HSAHR223200ug
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
HRP-Human Serum Albumin Protein, His Tag (SPR verified)
Serum albumin (SA) is also known as ALB, which is the main protein of plasma and has a good binding capacity for water,Ca2+,Na+,K+,fatty acids,hormones, bilirubin and drugs.The main function of SA is the regulation of the colloidal osmotic pressure of blood. As Major zinc transporter in plasma, SA typically binds about 80% of all plasma zinc. A variant structure of albumin could lead to increased binding of zinc resulting in an asymptomatic augmentation of zinc concentration in the blood. Defects in serum albumin can cause familial dysalbuminemic hyperthyroxinemia which is a form of euthyroid hyperthyroxinemia that is due to increased affinity of serum albumin for T4. It is the most common cause of inherited euthyroid hyperthyroxinemia in Caucasian population.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 200 μg
Product Type Albumin Protein