Learn More
Sperian Protection™ Honeywell™ Spacel Coverall
Brand:  Sperian Protection™ 4503000-M
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15209968
  • 4380.86 NOK / Pack of 25
Estimated Shipment
18-08-2020
Add to basket
Specifications
Specifications
Elastic
Yellow
Chemistry, petrochemistry, in-shore/off-shore oil industry, iIndustrial maintenance, pharmaceutical industry, spoiled beach cleaning, transportation of hazardous materials, iIndustrial sub-contracting cleaning, sanitation, waste processing, emergency services.
Spacel Coverall
100 μm
Polyethylene Film
Yes
Unisex
Medium